KEY001129489 Key Organics / BIONET

(2E)-3-[2-(4-methoxyphenoxy)phenyl]prop-2-enoic acid

  • MDL number MFCD01568763
  • Cas Number : 1164549-12-5
  • Molecular Weight : 270.284

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 208 - 210
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/8999

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/12N-036_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart