KEY001129492 Key Organics / BIONET

(2E)-3-[2-(4-methylphenoxy)phenyl]prop-2-enoic acid

  • MDL number MFCD01568766
  • Cas Number : 1164477-18-2
  • Molecular Weight : 254.285

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 213 - 215
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/9002

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/12N-040_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart