KEY001129520 Key Organics / BIONET

1-{2-[(2-chloro-1,3-thiazol-5-yl)methoxy]phenyl}ethan-1-one

  • MDL number MFCD01568772
  • Cas Number : 320423-63-0
  • Molecular Weight : 267.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 65 - 67
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/9029

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/12N-077_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart