KEY001129549 Key Organics / BIONET

4'-cyano-[1,1'-biphenyl]-4-carboxylic acid

  • MDL number MFCD00438676
  • Cas Number : 5728-46-1
  • Molecular Weight : 223.231

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 263 - 266
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/9058

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/12N-132_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart