KEY001129557 Key Organics / BIONET

4-(2-hydroxyethoxy)benzoic acid

  • MDL number MFCD00020367
  • Cas Number : 1711-24-6
  • Molecular Weight : 182.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 177 - 179
Iupac Name 4-(2-hydroxyethoxy)benzoic acid
Inchi Code 1S/C9H10O4/c10-5-6-13-8-3-1-7(2-4-8)9(11)12/h1-4,10H,5-6H2,(H,11,12)
Inchi Key QLIQIXIBZLTPGQ-UHFFFAOYSA-N

Description

Melting Point 177 - 179
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/9066

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/12N-142_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart