KEY001129678 Key Organics / BIONET

methyl 4H,5H-naphtho[1,2-b]thiophene-2-carboxylate

  • MDL number MFCD01569124
  • Cas Number : 320423-96-9
  • Molecular Weight : 244.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 85 - 88
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/9193

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/12N-502S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart