KEY001129682 Key Organics / BIONET

4H,5H-naphtho[1,2-b]thiophene-2-carboxylic acid

  • MDL number MFCD01569125
  • Cas Number : 29179-41-7
  • Molecular Weight : 230.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 227 - 230
Iupac Name 4,5-dihydronaphtho[1,2-b]thiophene-2-carboxylic acid
Inchi Code 1S/C13H10O2S/c14-13(15)11-7-9-6-5-8-3-1-2-4-10(8)12(9)16-11/h1-4,7H,5-6H2,(H,14,15)
Inchi Key IGBRCZHGFVMFCR-UHFFFAOYSA-N

Description

Melting Point 227 - 230
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/9197

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/12N-508S_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart