KEY001129878 Key Organics / BIONET

[4-(3-fluoropropoxy)phenyl]methanol

  • MDL number MFCD02091005
  • Cas Number : 449778-58-9
  • Molecular Weight : 184.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 52 - 54
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/9412

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/12P-148_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart