KEY001130004 Key Organics / BIONET

1-(1H-1,3-benzodiazol-2-yl)ethan-1-one

  • MDL number MFCD00159994
  • Cas Number : 939-70-8
  • Molecular Weight : 160.176

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 190 - 192
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/9556

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/12P-660_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart