KEY001130049 Key Organics / BIONET

N4,N4-dimethyl-5-phenylpyrimidine-2,4-diamine

  • MDL number MFCD00127225
  • Cas Number : 320424-61-1
  • Molecular Weight : 214.272

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 197 - 199
Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/63168

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1B-058_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart