KEY001130093 Key Organics / BIONET

methyl 3-(2-chlorophenyl)-5-methyl-1,2-oxazole-4-carboxylate

  • MDL number MFCD00214711
  • Cas Number : 4357-94-2
  • Molecular Weight : 251.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 56 - 58
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/63215

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1C-077_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart