KEY001130241 Key Organics / BIONET

1-[4-(2-chloroethyl)phenyl]ethan-1-one

  • MDL number MFCD00016350
  • Cas Number : 69614-95-5
  • Molecular Weight : 182.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 106@0.2mm Hg
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/63368

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1D-150_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart