KEY001130419 Key Organics / BIONET

4-(2,4-dichlorobenzoyl)-1-methyl-1H-pyrrole-2-carboxylic acid

  • MDL number MFCD00173071
  • Cas Number : 320419-40-7
  • Molecular Weight : 298.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 206 - 208
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/63565

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1F-048_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart