KEY001130448 Key Organics / BIONET

1-[(2-chlorophenyl)methyl]-2-oxo-1,2-dihydropyridine-3-carboxylic acid

  • MDL number MFCD00140815
  • Cas Number : 66158-19-8
  • Molecular Weight : 263.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 170 - 174
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/63597

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1F-318S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart