KEY001130455 Key Organics / BIONET

2-oxo-1-(prop-2-en-1-yl)-1,2-dihydropyridine-3-carboxylic acid

  • MDL number MFCD00140660
  • Cas Number : 66158-33-6
  • Molecular Weight : 179.175

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 126 - 127
Purity 95
Storage temp. RT
Physical Form White to Off-white Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/63614

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/1F-340S_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart