KEY001130483 Key Organics / BIONET

2,4-dioxo-1-(piperidin-1-yl)-1,2,3,4-tetrahydropyrimidine-5-carbonitrile

  • MDL number MFCD00172932
  • Cas Number : 320420-05-1
  • Molecular Weight : 220.232

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 265 - 269
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/63653

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1F-903_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart