KEY001130492 Key Organics / BIONET

5H,6H-benzo[h]quinazolin-2-amine

  • MDL number MFCD00139519
  • Cas Number : 66521-84-4
  • Molecular Weight : 197.241

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 193 - 196
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/63663

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1F-915_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart