KEY001130504 Key Organics / BIONET

N-(4-aminophenyl)-2-(phenylformamido)acetamide

  • MDL number MFCD00172388
  • Cas Number : 108717-59-5
  • Molecular Weight : 269.304

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 228 - 230
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/63678

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1F-946_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart