KEY001130686 Key Organics / BIONET

methyl 2-amino-6-[3-chloro-5-(trifluoromethyl)pyridin-2-yl]-4H,5H,6H,7H-thieno[2,3-c]pyridine-3-carboxylate

  • MDL number MFCD00180791
  • Cas Number : 321430-29-9
  • Molecular Weight : 391.79

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 149 - 151
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/63881

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1G-907_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart