KEY001130704 Key Organics / BIONET

2-amino-6-[3-chloro-5-(trifluoromethyl)pyridin-2-yl]-4H,5H,6H,7H-thieno[2,3-c]pyridine-3-carbonitrile

  • MDL number MFCD00180796
  • Cas Number : 321430-41-5
  • Molecular Weight : 358.77

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 199 - 202
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/63899

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1G-929_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart