KEY001130716 Key Organics / BIONET

ethyl (2Z)-2-(hydroxyimino)-3-oxobutanoate

  • MDL number MFCD00039517
  • Cas Number : 66508-93-8
  • Molecular Weight : 159.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 46 - 48
Iupac Name ethyl 2-nitroso-3-oxobutanoate
Inchi Code 1S/C6H9NO4/c1-3-11-6(9)5(7-10)4(2)8/h5H,3H2,1-2H3
Inchi Key FZFHKHNJQOFKAT-UHFFFAOYSA-N

Description

Melting Point 46 - 48
Purity 95%
Storage temp. Store under N2 at -18
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/63912

Safety Information

signalword Warning
pcodes P411,P422, P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/1G-944_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart