KEY001130762 Key Organics / BIONET

{1-[3-chloro-5-(trifluoromethyl)pyridin-2-yl]-1H-pyrrol-2-yl}methanol

  • MDL number MFCD00231916
  • Cas Number : 321430-86-8
  • Molecular Weight : 276.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 96 - 98
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/63960

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1H-049_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart