KEY001130864 Key Organics / BIONET

2-[(2,4-dichlorophenyl)methoxy]naphthalene-1-carbaldehyde

  • MDL number MFCD00202630
  • Cas Number : 321432-08-0
  • Molecular Weight : 331.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 178 - 180
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/64071

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1H-918_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart