KEY001130888 Key Organics / BIONET

ethyl 5-{[(1-formylnaphthalen-2-yl)oxy]methyl}-4,5-dihydro-1,2-oxazole-3-carboxylate

  • MDL number MFCD00665159
  • Cas Number : 321432-25-1
  • Molecular Weight : 327.336

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 113 - 115
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/64098

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1H-950_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart