KEY001131118 Key Organics / BIONET

6-hydroxy-1-methyl-2-oxo-1,2-dihydropyrimidine-4-carboxylic acid

  • MDL number MFCD00244509
  • Cas Number : 439109-81-6
  • Molecular Weight : 170.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 316 - 320
Iupac Name 1-methyl-2,6-dioxo-1,2,3,6-tetrahydro-4-pyrimidinecarboxylic acid
Inchi Code 1S/C6H6N2O4/c1-8-4(9)2-3(5(10)11)7-6(8)12/h2H,1H3,(H,7,12)(H,10,11)
Inchi Key OEGWVZNGNUHUQP-UHFFFAOYSA-N

Description

Melting Point 316 - 320
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/64387

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/1K-522S_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart