KEY001131408 Key Organics / BIONET

5-chloro-1-methyl-3-{[(4-methylphenyl)sulfanyl]methyl}-1H-pyrazole-4-carbaldehyde

  • MDL number MFCD00794830
  • Cas Number : 321526-27-6
  • Molecular Weight : 280.77

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 93 - 95.5
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/64702

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1L-644S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart