KEY001131413 Key Organics / BIONET

4-chloro-2-(3-chlorophenyl)-5-(piperazin-1-yl)-2,3-dihydropyridazin-3-one

  • MDL number MFCD00793826
  • Cas Number : 449778-66-9
  • Molecular Weight : 325.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 131 - 133
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/64710

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1L-905_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart