KEY001131442 Key Organics / BIONET

ethyl 2-[(thiophen-2-yl)formamido]acetate

  • MDL number MFCD00701883
  • Cas Number : 39978-25-1
  • Molecular Weight : 213.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 89 - 91
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/64739

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1M-012_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart