KEY001131453 Key Organics / BIONET

5-ethoxy-2-(4-methoxyphenyl)-1,3-oxazole

  • MDL number MFCD00793751
  • Cas Number : 477867-59-7
  • Molecular Weight : 219.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 47 - 49
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/64750

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1M-023_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart