KEY001131502 Key Organics / BIONET

1-(5-chloro-1-methyl-3-phenyl-1H-pyrazol-4-yl)ethan-1-one

  • MDL number MFCD01314892
  • Cas Number : 321526-28-7
  • Molecular Weight : 234.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 109 - 111
Purity 92
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/64803

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1M-504S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart