KEY001131541 Key Organics / BIONET

1-(3-bromophenyl)-3,3-dimethylazetidin-2-one

  • MDL number MFCD00793408
  • Cas Number : 338392-04-4
  • Molecular Weight : 254.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 61 - 63
Iupac Name 1-(3-bromophenyl)-3,3-dimethyl-2-azetidinone
Inchi Code 1S/C11H12BrNO/c1-11(2)7-13(10(11)14)9-5-3-4-8(12)6-9/h3-6H,7H2,1-2H3
Inchi Key ZOVGIJCGWKYQCT-UHFFFAOYSA-N

Description

Melting Point 61 - 63
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/64845

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/1M-702_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart