KEY001131579 Key Organics / BIONET

ethyl 2-{3-[(4-fluorophenyl)carbamoyl]pyridin-2-yl}acetate

  • MDL number MFCD00974991
  • Cas Number : 338392-11-3
  • Molecular Weight : 302.305

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 91 - 93
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/64889

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1M-908_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart