KEY001131602 Key Organics / BIONET

ethyl 6-oxo-2-(trifluoromethyl)-1,4,5,6-tetrahydropyridine-3-carboxylate

  • MDL number MFCD00975005
  • Cas Number : 194673-12-6
  • Molecular Weight : 237.178

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 128 - 130
Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/64912

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1M-938_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart