KEY001131688 Key Organics / BIONET

5-(2-hydroxyethyl)-6-methyl-2-sulfanyl-3,4-dihydropyrimidin-4-one

  • MDL number MFCD01316114
  • Cas Number : 21585-16-0
  • Molecular Weight : 186.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 270
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/65026

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1N-732_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart