KEY001131698 Key Organics / BIONET

methyl 3-[(2-chloro-1,3-thiazol-5-yl)methoxy]benzoate

  • MDL number MFCD01568782
  • Cas Number : 338393-42-3
  • Molecular Weight : 283.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 97 - 99
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/65037

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1P-003_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart