KEY001131705 Key Organics / BIONET

2-[(2-chloro-1,3-thiazol-5-yl)methoxy]benzaldehyde

  • MDL number MFCD01568787
  • Cas Number : 338393-47-8
  • Molecular Weight : 253.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 75 - 77
Iupac Name 2-[(2-chloro-1,3-thiazol-5-yl)methoxy]benzaldehyde
Inchi Code 1S/C11H8ClNO2S/c12-11-13-5-9(16-11)7-15-10-4-2-1-3-8(10)6-14/h1-6H,7H2
Inchi Key NSJLQTQYCXUGPQ-UHFFFAOYSA-N

Description

Melting Point 75 - 77
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/65045

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/1P-012_SDS.pdf
hcodes H302,H312,H315,H319,H332,H335
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart