KEY001131719 Key Organics / BIONET

2-(3,4-difluorophenoxy)benzaldehyde

  • MDL number MFCD01568793
  • Cas Number : 289717-89-1
  • Molecular Weight : 234.202

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 61 - 64 / 110 - 112@ 0.2mm Hg
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/65060

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1P-030_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart