KEY001131770 Key Organics / BIONET

[(E)-(1-phenylethylidene)amino]urea

  • MDL number MFCD00137159
  • Cas Number : 2492-30-0
  • Molecular Weight : 177.207

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 207 - 209
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/65115

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1P-098_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart