KEY001132093 Key Organics / BIONET

3-[3-(2,5-dimethyl-1H-pyrrol-1-yl)thiophen-2-yl]-3-oxopropanenitrile

  • MDL number MFCD02102411
  • Cas Number : 477871-28-6
  • Molecular Weight : 244.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 156.5 - 158
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/65461

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1R-0233_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart