KEY001132161 Key Organics / BIONET

1-(3,4-dichlorophenyl)-3,5-dimethyl-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD02102423
  • Cas Number : 477710-54-6
  • Molecular Weight : 285.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 238 - 240
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/65528

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1R-0688_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart