KEY001132290 Key Organics / BIONET

1-(4-chlorobenzenesulfonyl)piperidine-4-carbohydrazide

  • MDL number MFCD00754333
  • Cas Number : 332400-85-8
  • Molecular Weight : 317.79

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 206 - 208
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/65678

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1R-1300_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart