KEY001132442 Key Organics / BIONET

quinoxaline-2-thiol

  • MDL number MFCD00138789
  • Cas Number : 6962-54-5
  • Molecular Weight : 162.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 192 - 195
Iupac Name 2-quinoxalinethiol
Inchi Code 1S/C8H6N2S/c11-8-5-9-6-3-1-2-4-7(6)10-8/h1-5H,(H,10,11)
Inchi Key INQXGZFDGDSRIF-UHFFFAOYSA-N

Description

Melting Point 192 - 195
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/66709

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/2D-065_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart