KEY001132568 Key Organics / BIONET

methyl 4-hydroxy-1-(4-methoxyphenyl)-6-oxo-1,6-dihydropyridazine-3-carboxylate

  • MDL number MFCD00141150
  • Cas Number : 121582-61-4
  • Molecular Weight : 276.248

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 184 - 186
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/66850

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/2F-041_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart