KEY001132580 Key Organics / BIONET

2-[1-methyl-5-(2,2,2-trichloroacetyl)-1H-pyrrole-3-carbonyl]benzoic acid

  • MDL number MFCD00141858
  • Cas Number : 338395-82-7
  • Molecular Weight : 374.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 214 - 216
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/66864

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/2F-055_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart