KEY001132598 Key Organics / BIONET

ethyl [(4-fluorophenyl)carbamoyl]formate

  • MDL number MFCD00119045
  • Cas Number : 69065-91-4
  • Molecular Weight : 211.192

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 120 - 122
Purity 95
Storage temp. RT
Physical Form White to Off-white Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/66884

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/2F-079_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart