KEY001132603 Key Organics / BIONET

1-(hydrazinecarbonyl)-N-(4-methoxyphenyl)formamide

  • MDL number MFCD00173090
  • Cas Number : 20580-47-6
  • Molecular Weight : 209.205

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 236 - 238
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/66889

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/2F-084_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart