KEY001132627 Key Organics / BIONET

6-(1H-pyrazol-1-yl)pyridine-3-carboxylic acid

  • MDL number MFCD00140749
  • Cas Number : 253315-22-9
  • Molecular Weight : 189.174

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 272 - 274
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/66921

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/2F-330S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart