KEY001132662 Key Organics / BIONET

1-methyl-5-(phenylsulfanyl)-3-(trifluoromethyl)-1H-pyrazole-4-carbaldehyde

  • MDL number MFCD00140823
  • Cas Number : 321533-77-1
  • Molecular Weight : 286.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 96
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/66968

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/2F-393S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart