KEY001132686 Key Organics / BIONET

5-{[(2-chlorophenyl)methyl]sulfanyl}-1-methyl-3-(trifluoromethyl)-1H-pyrazole-4-carbaldehyde

  • MDL number MFCD00140828
  • Cas Number : 321533-90-8
  • Molecular Weight : 334.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 86 - 88
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/66992

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/2F-417S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart