KEY001132697 Key Organics / BIONET

ethyl 5-phenyl-2-(pyrrolidin-1-yl)-1H-pyrrole-3-carboxylate

  • MDL number MFCD00171771
  • Cas Number : 118082-59-0
  • Molecular Weight : 284.359

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 167 - 169
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/67005

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/2F-905_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart